Anti-ABCC5 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIM4688-27FEA 587 EUR
USBIM4688-27F
Anti-ABCC5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: ABCC5 (ATP-binding cassette, sub-family C (CFTR/MRP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR