Anti-PTPN1 Goat Polyclonal Antibody

Supplier: United States Biological
USBIP9109-03EA 783 EUR
USBIP9109-03
Anti-PTPN1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR