Anti-Secretoneurin Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS0626-12AEA 979 EUR
USBIS0626-12A
Anti-Secretoneurin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Secretoneurin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR