Anti-SPRY1 Mouse Monoclonal Antibody [clone: 3H4]

Supplier: United States Biological
USBIS5620-01FEA 703 EUR
USBIS5620-01F
Anti-SPRY1 Mouse Monoclonal Antibody [clone: 3H4]
Antibodies
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SPRY1 (sprouty homolog 1, antagonist of FGF signaling (Drosophila))
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR