Anti-Alpha-1-Antitrypsin Goat Polyclonal Antibody
Supplier: Immunoreagents
IMMRIR1369EA 0 EUR
IMMRIR1369
Anti-Alpha-1-Antitrypsin Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Alpha-1-Antitrypsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human