Anti-IgG Goat Polyclonal Antibody

Supplier: Immunoreagents
IMMRIR1876EA 191 EUR
IMMRIR1876
Anti-IgG Goat Polyclonal Antibody
Antibodies
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifikace: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Informace pro objednávání: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Llama
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR