Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Supplier: Immunoreagents
IMMRIR1959EA 329 EUR
IMMRIR1959
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse