Anti-IgG Goat Polyclonal Antibody (DyLight® 594)

Supplier: Immunoreagents
IMMRIR2192EA 241 EUR
IMMRIR2192
Anti-IgG Goat Polyclonal Antibody (DyLight® 594)
Antibodies
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-colour applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593 nm/618 nm Emission Colour = Red (Similar Dyes: Alexa Fluor 594, Texas Red)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifikace: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Informace pro objednávání: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR