Printers for glassware machines
Supplier: Miele
141-0616EA 1035 EUR
141-0616 141-0630 141-0413
Printers for glassware machines
Printers Document Printers
The PRT 100 printer can be linked to a dishwasher via the RS232 module (included with 141-0413 and 141-0616) this is an ideal low-cost solution for small installations without a PC or for entry-level process documentation software packages.