Latex tests, Dryspot
Supplier: THERMO FISHER DIAGNOSTICS
DR0100M
DR0500G
DR0585A
DR0586G
DR0587G
DR0588G
DR0589G
DR0590G
DR0591G
DR0592G
DR0593G
DR0595A
DR0800M
DR0801M
DR0802M
DR0803M
DR0850M
DR0900A
DR0420M
DR0850B
DR0220M
DR1108A
DR0200M
DR0210M
OXOIDR0100MEA
249
EUR
OXOIDR0100M
OXOIDR0500G
OXOIDR0585A
OXOIDR0586G
OXOIDR0587G
OXOIDR0588G
OXOIDR0589G
OXOIDR0590G
OXOIDR0591G
OXOIDR0592G
OXOIDR0593G
OXOIDR0595A
OXOIDR0800M
OXOIDR0801M
OXOIDR0802M
OXOIDR0803M
OXOIDR0850M
OXOIDR0900A
OXOIDR0420M
OXOIDR0850B
OXOIDR0220M
OXOIDR1108A
OXOIDR0200M
OXOIDR0210M
Latex tests, Dryspot
Microbiology Test Systems
Antigen-Antibody Tests
Ag-AK, Latex
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
- Simplified room temperature storage allows storage for up to two years
- Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards