Chemical resistant overalls, 4570
Supplier: 3M
4570L 4570XL 4570M 45702XL 45703XL 45704XL 4570S
MMMA4570LEA 717 EUR
MMMA4570L MMMA4570XL MMMA4570M MMMA45702XL MMMA45703XL MMMA45704XL MMMA4570S
Chemical resistant overalls, 4570
Disposable Garments
These disposable overalls made of PP with PE film are designed to help protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4), and high pressure liquid jets (Type 3). They are also certified to offer protection against radioactive particulates and infective agents.
  • Elasticated hood with three panel design
  • Elasticated waist, wrists and ankles for convenience and freedom of movement
  • Two-way zipper with sealable storm flap and chin flap
  • Seams are taped with multi-layer clear tape
  • Anti-static treated (inside only)
Ideal for use in chemical industries, e.g. for decontamination, tank cleaning,‎ chemical mixing and handling, and pesticide spraying.
Colour: Grey
Certifikace: Type 3/4/5/6, EN 1073-2, EN 14126, EN 1149-5; Cat. III.