Filter tips, OneTouch™
Supplier: Sorenson Bioscience
10350 10320 10380 10510
732-3246EA 172 EUR
732-3246 732-3245 732-3247 732-3248
Filter tips, OneTouch™
Pipette Tips
Featuring a dual-shot tip design, these pipette tips combine virgin PP and pliable TPE to provide a rigid and dependable tip with a soft, easy-to-seal, color-coded collar. The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
  • Tips reduce the necessary sealing and reciprocal ejection force felt with traditional single resin PP tips
  • Minimal force required to eject tip
  • Ideal for multi-channel pipettors
  • Certified RNase/DNase-free
  • Top collar colour-coded by volume