Measuring cylinders, tall form, class B, borosilicate glass
Supplier: KAVALIER GLASS
632432141130 632432141444 632432141546 632432140923 632432140718 632432141125 632432141238 632432141343
KAVA632432141130EA 41.7 EUR
KAVA632432141130 KAVA632432141444 KAVA632432141546 KAVA632432140923 KAVA632432140718 KAVA632432141125 KAVA632432141238 KAVA632432141343
Measuring cylinders, tall form, class B, borosilicate glass
Cylinders Measuring Cylinders
Borosilicate glass, tall form, class B.
  • Hexagonal base and spout
  • White graduations
Certifikace: ISO 4788, DIN 12680