Anti- IgG Antibody (Unconjugated)

Supplier: Boster
BSBTBA1043-1EA 0 EUR
BSBTBA1043-1
Anti- IgG Antibody (Unconjugated)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR