Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))

Supplier: Boster
BSBTBA1070-0.5EA 0 EUR
BSBTBA1070-0.5
Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR