Anti-Avidin Antibody (Peroxidase)

Supplier: Boster
BSBTBA1081-1EA 0 EUR
BSBTBA1081-1
Anti-Avidin Antibody (Peroxidase)
Antibodies
Type: Primary
Antigen: Avidin
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR