Anti-IgG Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Boster
BSBTBA1089-0.5EA 0 EUR
BSBTBA1089-0.5
Anti-IgG Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR