Anti-CD63 Mouse Monoclonal Antibody [clone: AD1]

Supplier: United States Biological
USBIC2418-02L-100EA 561 EUR
USBIC2418-02L-100 USBIC2418-02L-25
Anti-CD63 Mouse Monoclonal Antibody [clone: AD1]
Antibodies
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: AD1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR