Anti-Bromodeoxyuridine Mouse Monoclonal Antibody [clone: BRD469]

Supplier: United States Biological
USBI138500EA 270 EUR
USBI138500
Anti-Bromodeoxyuridine Mouse Monoclonal Antibody [clone: BRD469]
Antibodies
Recommended dilution: Immunofluorescence: 1:50-1:100; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: Bromodeoxyuridine
Clonality: Monoclonal
Clone: BRD469
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR