Anti-Tissue Inhibitor of Metalloproteinases 3 Rabbit Polyclonal Antibody

Supplier: US Biological
138599 138599-100
USBI138599EA 391 EUR
USBI138599 USBI138599-100
Anti-Tissue Inhibitor of Metalloproteinases 3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ; Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunochemistry: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g

Type: Primary
Antigen: Tissue Inhibitor of Metalloproteinases 3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Horse, Human, Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR