Anti-4E Binding Protein 2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI0004-30-100EA 668 EUR
USBI0004-30-100
Anti-4E Binding Protein 2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 4E Binding Protein 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rabbit, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR