Anti-SMAD3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK600-401-919SEA 219 EUR
ROCK600-401-919S
Anti-SMAD3 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, immunochemistry and by western blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:75000 - 1:300,000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:500;

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Xenopus laevis, Xenopus Tropicalis, Zebrafish, Rat, Mouse, Pig, Bovine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR