Anti-Theophylline Mouse Monoclonal Antibody

Supplier: FITZGERALD INDUSTRIES INT.
FITZ10-T10-M28163EA 0 EUR
FITZ10-T10-M28163
Anti-Theophylline Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Theophylline
Clonality: Monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR