Anti-ULK1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP3867AEA 348 EUR
ABGEAP3867A
Anti-ULK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ULK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR