Anti-NR2F6 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP16861AEA 348 EUR
ABGEAP16861A
Anti-NR2F6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NR2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR