Anti-Meloxicam Polyclonal Antibody

Supplier: Abnova
ABNOPAB8301EA 1160 EUR
ABNOPAB8301
Anti-Meloxicam Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against Meloxicam.

Recommended Dilutions: Enzyme Immunoassay (0.156 ?g/ml

Type: Primary
Antigen: Meloxicam
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR