Anti-LCK Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB14161EA 419 EUR
ABNOPAB14161
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of LCK.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR