Anti-HSV1 Mouse Monoclonal Antibody

Supplier: Abnova
ABNOMAB6169EA 399 EUR
ABNOMAB6169
Anti-HSV1 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against herpes simplex virus type 1.

Type: Primary
Antigen: HSV1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR