Anti-APOB Goat Polyclonal Antibody

Supplier: Abnova
ABNOPAB13886EA 399 EUR
ABNOPAB13886
Anti-APOB Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against APOB.

Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR