Anti-IGFBP3 Mouse Monoclonal Antibody [clone: SBH10]

Supplier: Abnova
ABNOMAB7879EA 419 EUR
ABNOMAB7879
Anti-IGFBP3 Mouse Monoclonal Antibody [clone: SBH10]
Antibodies
Mouse monoclonal antibody raised against recombinant IGFBP3.

Recommended Dilutions: ELISA: 0.5-1 ?g/ml; Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: IGFBP3
Clonality: Monoclonal
Clone: SBH10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR