Anti-ELANE Mouse Monoclonal Antibody

Supplier: Abnova
ABNOMAB2202EA 399 EUR
ABNOMAB2202
Anti-ELANE Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against native ELANE.

Type: Primary
Antigen: ELANE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR