Anti-FOXO1 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB10970EA 419 EUR
ABNOPAB10970
Anti-FOXO1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of FOXO1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000

Type: Primary
Antigen: FOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR