Anti-CASP3 Polyclonal Antibody

Supplier: Abnova
ABNOPAB13561EA 1030 EUR
ABNOPAB13561
Anti-CASP3 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant CASP3.

Recommended Dilutions: Western Blot: 2-10 ?g/ml

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR