Anti-EIF4EBP1 Mouse Monoclonal Antibody [clone: 11G12C11]

Supplier: Abnova
ABNOMAB10437EA 419 EUR
ABNOMAB10437
Anti-EIF4EBP1 Mouse Monoclonal Antibody [clone: 11G12C11]
Antibodies
Mouse monoclonal antibody raised against recombinant EIF4EBP1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: EIF4EBP1
Clonality: Monoclonal
Clone: 11G12C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR