Anti-RAB2A Mouse Monoclonal Antibody

Supplier: Abnova
ABNOMAB11163EA 399 EUR
ABNOMAB11163
Anti-RAB2A Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against partial recombinant protein of RAB2A.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:100-1:2000

Type: Primary
Antigen: RAB2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR