Anti-CHEK1 Mouse Monoclonal Antibody [clone: 2G1D5]
Supplier: Abnova
ABNOMAB10554EA 399 EUR
ABNOMAB10554
Anti-CHEK1 Mouse Monoclonal Antibody [clone: 2G1D5]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CHEK1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: CHEK1
Clonality: Monoclonal
Clone: 2G1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse