Bacterial antigen rapid latex agglutination test, Wellcogen™
Supplier: THERMO FISHER DIAGNOSTICS
R30858701
R30858801
R30859001
R30859203
R30859502
R30859602
R30164501
R30368601
OXOIR30858701EA
306
EUR
OXOIR30858701
OXOIR30858801
OXOIR30859001
OXOIR30859203
OXOIR30859502
OXOIR30859602
OXOIR30164501
OXOIR30368601
Bacterial antigen rapid latex agglutination test, Wellcogen™
Microbiology Test Systems
Antigen-Antibody Tests
Ag-AK, Latex
Directly detect antigens present in human cerebrospinal fluid (CSF), serum, urine, or blood cultures using Thermo Scientific™ Wellcogen™ Bacterial Antigen Rapid Latex Agglutination Test.
- Not all products are available for sale in all territories