Pycnometers, Hubbard
Supplier: NEUBERT VOLUM. GLASSWAERE
0303-50-24 0303-50-25 0303-50-50
NEUB0303-50-24EA 102 EUR
NEUB0303-50-24 NEUB0303-50-25 NEUB0303-50-50
Pycnometers, Hubbard
Specific Gravity Bottles
Borosilicate glass 3,3.
  • With glass stopper
  • For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.