ESD safety shoes, lace-up, Senex Pro GTX
Supplier: ELTEN
728571/39 728571/38 728571/40 728571/41 728571/42 728571/43 728571/44 728571/45 728571/46 728571/47 728571/48 728571/36 728571/37
114-6247EA 112.9 EUR
114-6247 114-6246 114-6248 114-6249 114-6250 114-6251 114-6252 114-6253 114-6254 114-6255 114-8224 ELTN728571/36 ELTN728571/37
ESD safety shoes, lace-up, Senex Pro GTX
Shoes and Boots
100% metal- and leather-free, sporty, lightweight ESD safety shoes made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water-repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Gore-Tex® climate membrane
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.