Calibration gas cylinders
Supplier: MSA
111-1703EA 844 EUR
111-1703
Calibration gas cylinders
Standards Petroleum Industry Standards
The gas mixture in calibration gas cylinder is certified to be prepared gravimetrically, using NIST traceable weights. The lot number and nominal value of the gas constituents in volume, percent by mass, PPM, or volume is specified on the cylinder.