Gas detection tubes for mercaptans
Supplier: Dräger
6728981 8101871 8103281
151-0160EA 114 EUR
151-0160 151-0182 118-0322
Gas detection tubes for mercaptans
Gas Tubes
To measure the 'spot concentrations' of mercaptan for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Order Now
SPECIFICATIONS
Model Mercaptan 0.5/a Mercaptan 20/a Mercaptan 0.1/a
Environmentally Preferable
Colour change White to yellow White to yellow brown Yellow to red
Humidity 3 - 15 mg/l H₂O 3 - 30 mg/l H₂O 2 - 40 mg/l H₂O
Measurement time 8 minutes 2,5 minutes 3 minutes/40 seconds
No. of strokes 20 10 10 / 2
Standard deviation ±10 - 15 % ±10 - 15 % ±10 - 15 %
Standard range 0,5 - 5 ppm 2 - 100 ppm 0,1 - 2,5 / 3 - 15 ppm
Temperature range 10…40 °C 0…50 °C 5…40 °C