Chemical resistant ESD gloves, uvex rubiflex NB35A
Supplier: UVEX
60954/06 60954/09 60954/10 60954/11 60954/07 60954/08
112-4398EA 201 EUR
112-4398 112-4410 112-4411 112-4412 UVEX60954/07 UVEX60954/08
Chemical resistant ESD gloves, uvex rubiflex NB35A
Gloves
These extremely light and flexible chemical protection gloves feature a cotton interlock/carbon lining and a full nitrile butadiene rubber (NBR) coating.
  • Good resistance to grease, mineral oils and many chemicals
  • Good mechanical properties
  • Anti-static in accordance with EN 16350
  • High water vapour absorption due to the cotton lining
  • High flexibility, lightweight
  • Forward resistance of <10⁸ Ohm
Ideal for use in environments where ESD and chemical protection is required, for example in paint shops, in the chemical industry and in the automotive industry. Also suitable for use in areas where explosions are a risk.
Certifikace: EN 16350, EN ISO 374-1:2016 Type A (JKLOPT), EN 388:2016 (2111X)
OEKO-Tex® Standard 100