Markers, bel-pen, SP Bel-Art
Supplier: Bel-Art
BELAF133740000EA 55.4 EUR
BELAF133740000
Markers, bel-pen, SP Bel-Art
Pens Permanent Markers
These felt tip marking pens are perfect for labeling all types of glassware and plastics. The permanent and quick drying ink remains waterproof and smudge proof.
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 133 mm long