Sterilsop® SX removable heat sterilization bags
Supplier: Hartmann
Sterilsop®
HART6555051EA 0 EUR
HART6555051 HART6055232
Sterilsop® SX removable heat sterilization bags
Bags Sterilisation Pouches
Heat-sealable sterilization packages specially designed for wet heat sterilisation. Sterilsop® SX sachets are peelable flat bags and are intended for use as sterile barrier systems for the packaging of medical linen, medical and surgical equipment, dressing supplies.
  • Permeability to the sterilising agent
  • Maintenance of sterility of content
  • Aseptic extraction of sterilised material
  • One side paper with sterilisation indicators
  • A complex pink transparent polyester / polypropylene face
  • Free of latex, phthalates and rosin
Sterilsop® SX is a Class I device.
Shelf life: five years
Certifikace: Complies with EN ISO 11607-1 and EN 868-5 standards.