Underpads, MoliNea® Plus D
Supplier: Hartmann
MoliNea® Plus D
HART163700/9EA 0 EUR
HART163700/9
Underpads, MoliNea® Plus D
Surface Protectors Patient Underpads
These cellulose underpads are suitable for the protection of bed or chair. It is a medical device of the class.
  • Waterproof, non-slip foil backing
  • Absorbent core consisting of 20 layers of cellulose
  • Non-woven top layer covers the entire surface including the foil edges
  • Sealed completely from edge to edge