Capillary columns, Rtx®-50 (fused silica)
Supplier: Restek
Rtx®
REST10522EA 510 EUR
REST10522 REST10567 REST10523 REST10520 REST10542 REST10543 REST10521 REST10526 REST10527 REST10524 REST10569 REST40510 REST40511 REST10570 REST10551 REST10573 REST10552 REST10550 REST10555 REST10556 REST10553 REST10554 REST10537 REST10538 REST10557 REST10535 REST10558 REST10536 REST40501 REST10539 REST40502 REST10540 REST10541
Capillary columns, Rtx®-50 (fused silica)
Chromatography Columns
General purpose columns for pesticides, herbicides, rosin acids, phthalate esters, sterols. The high thermal stability of Rtx®-50 columns makes dual column analysis possible with common phases such as Rtx®-1 or Rtx®-5MS.
Midpolarity phase; Crossbond® phenyl methyl polysiloxane.
Temperature range: 40 to 320 °C
Similar phases: HP-50+, CP-Sil 24 CB, SPB-50, AT-50, 007-17
Equivalent to USP G3 phase.