Rotary evaporator splash head
Supplier: Smith Scientific
SMIT8SH1524EA 221 EUR
SMIT8SH1524
Rotary evaporator splash head
Distillation Components Distillation Heads
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes