Capillary columns, Rtx®-5Sil MS (fused silica)
Supplier: Restek
12705 12708 12721 12723 12724 12726 12736 12737 12738 12739 12740 12753 12754 12794 42702 42703 42704
REST12705EA 0 EUR
REST12705 REST12708 REST12721 REST12723 REST12724 REST12726 REST12736 REST12737 REST12738 REST12739 REST12740 REST12753 REST12754 REST12794 REST42702 REST42703 REST42704
Capillary columns, Rtx®-5Sil MS (fused silica)
Chromatography Columns
Rtx®-5Sil MS columns are optimised for use with conventional MS detectors but for the fastest analyses, they are equally compatible with time-of-flight mass spectrometers.
Crossbond®, selectivity similar to 5% diphenyl/95% dimethyl polysiloxane.
Temperature range: −60 °C to 330/350 °C