VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Supplier: VWR
VWR®
525-0923EA 67.9 EUR
525-0923
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Closures Test/Sample Tube Closures
Purple PP. Sustainable design based on virgin resins.
Closures for use with VWR® 50 ml tubes. Non sterile. Not autoclavable.