ESD safety winter boots, Ergo-Med AB 846 XP
Supplier: ATLAS - the shoe company
52114S3_W14_42 52114S3_W14_43 52113S3_W13_36 52113S3_W13_37 52113S3_W13_38 52113S3_W13_39 52113S3_W13_40 52113S3_W13_41 52113S3_W13_42 52113S3_W13_43 52113S3_W13_44 52113S3_W13_45 52113S3_W13_46 52113S3_W13_47 52113S3_W13_48 52113S3_W13_49 52113S3_W13_50 52114S3_W14_36 52114S3_W14_37 52114S3_W14_38 52114S3_W14_39 52114S3_W14_40 52114S3_W14_41 52114S3_W14_44 52114S3_W14_45 52114S3_W14_46 52114S3_W14_47 52114S3_W14_48 52114S3_W14_49 52114S3_W14_50
114-6886EA 269 EUR
114-6886 114-6887 114-6865 114-6866 114-6867 114-6868 114-6869 114-6870 114-6871 114-6872 114-6873 114-6874 114-6875 114-6876 114-6877 114-6878 114-6879 114-6880 114-6881 114-6882 114-6883 114-6884 114-6885 114-6888 114-6889 114-6890 114-6891 114-6892 114-6893 114-6894
ESD safety winter boots, Ergo-Med AB 846 XP
Shoes and Boots
Extra wide ESD safety winter boot made from waterproof leather, reflectors and an outdoor sole (4,5 mm rough tread profile, oil, acid and petrol resistant, heat resistant up to 170 °C).
  • Warm woven fur lining
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Puncture resistant, metal-free XP® midsole
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/blue
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, CI, SRC